Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATGTATTTGTGGCTTAAACTC[G/T]TGGCATTTGGCTTTGCCTTTCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 151460 | ||||||||||||||||||||
Literature Links: |
PTPRC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPRC - protein tyrosine phosphatase, receptor type C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267798.1 | 184 | Missense Mutation | GTG,TTG | V,L 10 | NP_001254727.1 | |
NM_002838.4 | 184 | Missense Mutation | GTG,TTG | V,L 10 | NP_002829.3 | |
NM_080921.3 | 184 | Missense Mutation | GTG,TTG | V,L 10 | NP_563578.2 | |
XM_006711472.3 | 184 | Missense Mutation | GTG,TTG | V,L 10 | XP_006711535.1 | |
XM_006711473.3 | 184 | Missense Mutation | GTG,TTG | V,L 10 | XP_006711536.1 | |
XM_006711474.3 | 184 | Missense Mutation | GTG,TTG | V,L 10 | XP_006711537.1 |