Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGAAAATGTGTCTCCTGAATAA[A/C]CAGAGAGAGAGAAGCTGGAGAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608322 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF21B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF21B - kinesin family member 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252100.1 | 6992 | UTR 3 | NP_001239029.1 | |||
NM_001252102.1 | 6992 | UTR 3 | NP_001239031.1 | |||
NM_001252103.1 | 6992 | UTR 3 | NP_001239032.1 | |||
NM_017596.3 | 6992 | UTR 3 | NP_060066.2 | |||
XM_017000731.1 | 6992 | Intron | XP_016856220.1 | |||
XM_017000732.1 | 6992 | Intron | XP_016856221.1 |
MROH3P - maestro heat like repeat family member 3, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |