Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGACCCAGAAACCGAACCCACC[C/T]GGGTACCTGTGGATCCCTGTCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616597 MIM: 176889 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL8A - ADP ribosylation factor like GTPase 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256129.1 | Intron | NP_001243058.1 | ||||
NM_138795.3 | Intron | NP_620150.1 |
GPR37L1 - G protein-coupled receptor 37 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPN7 - protein tyrosine phosphatase, non-receptor type 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |