Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTGTTCCTCTCCCCTTCTTCT[C/T]TCTCAATTCAGGGCAGGCCCAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 159980 MIM: 603145 | ||||||||||||||||||||
Literature Links: |
MYOG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYOG - myogenin (myogenic factor 4) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002479.5 | 1206 | UTR 3 | NP_002470.2 |
PPFIA4 - PTPRF interacting protein alpha 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |