Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCTCCCTGCCTCCTACCACAGA[C/G]GCCAGAATCACCTGGACCGAGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3C2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIK3C2B - phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002646.3 | 4465 | UTR 3 | NP_002637.3 | |||
XM_005245257.2 | 4465 | UTR 3 | XP_005245314.1 | |||
XM_005245258.3 | 4465 | UTR 3 | XP_005245315.1 | |||
XM_011509630.2 | 4465 | UTR 3 | XP_011507932.1 | |||
XM_011509631.2 | 4465 | UTR 3 | XP_011507933.1 | |||
XM_011509633.2 | 4465 | Intron | XP_011507935.1 | |||
XM_017001473.1 | 4465 | UTR 3 | XP_016856962.1 | |||
XM_017001474.1 | 4465 | UTR 3 | XP_016856963.1 | |||
XM_017001475.1 | 4465 | UTR 3 | XP_016856964.1 |