Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTAAATTGCAACCTCCTATCAC[C/T]TCCTCCTCCCCATTGCTCTTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRCC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRCC - papillary renal cell carcinoma (translocation-associated) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005973.4 | Intron | NP_005964.3 | ||||
XM_005245313.2 | Intron | XP_005245370.1 | ||||
XM_005245314.1 | Intron | XP_005245371.1 | ||||
XM_011509726.1 | Intron | XP_011508028.1 |