Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGACCAGCAAAGCCTGGGCCCC[A/G]AGTTGGATTGCCAGGCGCCGTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616406 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6741 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6741 - microRNA 6741 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYCR2 - pyrroline-5-carboxylate reductase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271681.1 | 623 | Silent Mutation | CTC,CTT | L,L 131 | NP_001258610.1 | |
NM_013328.3 | 623 | Silent Mutation | CTC,CTT | L,L 205 | NP_037460.2 |