Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCAGGAGCCCAAAGTGTTCTCA[A/G]TCAAACCCACCCACCCACTTCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF5L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF5L - TATA-box binding protein associated factor 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025247.1 | 2671 | Intron | NP_001020418.1 | |||
NM_014409.3 | 2671 | UTR 3 | NP_055224.1 | |||
XM_005273099.4 | 2671 | UTR 3 | XP_005273156.1 | |||
XM_005273100.2 | 2671 | UTR 3 | XP_005273157.1 | |||
XM_011544162.1 | 2671 | UTR 3 | XP_011542464.1 | |||
XM_011544163.1 | 2671 | UTR 3 | XP_011542465.1 | |||
XM_011544164.2 | 2671 | UTR 3 | XP_011542466.1 | |||
XM_017001045.1 | 2671 | UTR 3 | XP_016856534.1 |