Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGCGGCGGTGGCCATCTGCCA[G/T]CTGTGCGACCGCACCCCGCCAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610584 | ||||||||||||||||||||
Literature Links: |
TRIM67 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM67 - tripartite motif containing 67 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004342.3 | 549 | Missense Mutation | CAG,CAT | Q,H 209 | NP_001004342.3 | |
NM_001300889.1 | 549 | Missense Mutation | CAG,CAT | Q,H 169 | NP_001287818.1 | |
XM_011544192.2 | 549 | Missense Mutation | CAG,CAT | Q,H 209 | XP_011542494.1 | |
XM_017001323.1 | 549 | Missense Mutation | CAG,CAT | Q,H 209 | XP_016856812.1 |