Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTCTGGGTCACTATATCTCCTA[C/T]CCTGAATGGGGGAAGATGGGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615283 MIM: 616086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC8 - exocyst complex component 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175876.4 | 3441 | UTR 3 | NP_787072.2 |
SPRTN - SprT-like N-terminal domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |