Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGCCGCCTCTCAGGTTGTTGTG[C/T]GGGGTGCAGATGAGGCATCTCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHA8 - EPH receptor A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011540969.1 | Intron | XP_011539271.1 | ||||
XM_011540970.1 | Intron | XP_011539272.1 | ||||
XM_011540972.1 | Intron | XP_011539274.1 | ||||
XM_011540973.1 | Intron | XP_011539275.1 | ||||
XM_011540975.2 | Intron | XP_011539277.1 |