Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGGGAGCTTAAGGCCACACTCC[A/C]AGGCAGGGTATCTGCTGATATCGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAZN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KAZN - kazrin, periplakin interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017999.2 | Intron | NP_001017999.1 | ||||
NM_001018000.3 | Intron | NP_001018000.1 | ||||
NM_001018001.2 | Intron | NP_001018001.1 | ||||
NM_015209.2 | Intron | NP_056024.1 | ||||
NM_201628.2 | Intron | NP_963922.2 | ||||
XM_005245795.4 | Intron | XP_005245852.1 | ||||
XM_011541074.2 | Intron | XP_011539376.1 | ||||
XM_011541075.2 | Intron | XP_011539377.1 | ||||
XM_011541076.2 | Intron | XP_011539378.1 | ||||
XM_011541077.2 | Intron | XP_011539379.1 | ||||
XM_011541080.2 | Intron | XP_011539382.1 | ||||
XM_011541081.2 | Intron | XP_011539383.1 | ||||
XM_017000768.1 | Intron | XP_016856257.1 | ||||
XM_017000769.1 | Intron | XP_016856258.1 | ||||
XM_017000770.1 | Intron | XP_016856259.1 | ||||
XM_017000771.1 | Intron | XP_016856260.1 | ||||
XM_017000772.1 | Intron | XP_016856261.1 |