Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAACAAAAACAAAAAAAAAACTT[A/T]ACGCTTACCATTGCTGCATATTGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604002 | ||||||||||||||||||||
Literature Links: |
PQLC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PQLC3 - PQ loop repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282710.1 | 5619 | Intron | NP_001269639.1 | |||
NM_001282711.1 | 5619 | Intron | NP_001269640.1 | |||
NM_001282712.1 | 5619 | Intron | NP_001269641.1 | |||
NM_152391.4 | 5619 | Intron | NP_689604.1 |
ROCK2 - Rho associated coiled-coil containing protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321643.1 | 5619 | UTR 3 | NP_001308572.1 | |||
NM_004850.4 | 5619 | UTR 3 | NP_004841.2 | |||
XM_005246190.3 | 5619 | UTR 3 | XP_005246247.1 | |||
XM_011510417.2 | 5619 | UTR 3 | XP_011508719.1 | |||
XM_017005378.1 | 5619 | Intron | XP_016860867.1 | |||
XM_017005379.1 | 5619 | UTR 3 | XP_016860868.1 |