Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTCATCAATATCAATTGCCAAA[C/T]TCAGTCTGAAAACATTTCACACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601756 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALNT3 - polypeptide N-acetylgalactosaminyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004482.3 | Intron | NP_004473.2 | ||||
XM_005246449.1 | Intron | XP_005246506.1 | ||||
XM_011510929.1 | Intron | XP_011509231.1 | ||||
XM_017003770.1 | Intron | XP_016859259.1 |