Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTAATGTAAGAACCAAAGAAA[A/T]CCTTAAGCCATAAAAGCCAGAAACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610643 MIM: 609929 | |||||||||||||||||||||||
Literature Links: |
KIAA1524 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KIAA1524 - KIAA1524 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020890.2 | 2995 | UTR 3 | NP_065941.2 | |||
XM_006713716.3 | 2995 | UTR 3 | XP_006713779.1 | |||
XM_006713717.2 | 2995 | UTR 3 | XP_006713780.1 | |||
XM_011513056.1 | 2995 | UTR 3 | XP_011511358.1 | |||
XM_011513057.2 | 2995 | UTR 3 | XP_011511359.1 |
MYH15 - myosin heavy chain 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |