Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGACTGGATGGCCTCGCTCCCC[C/G]CTCACCTCCACAACCTCCCCCTTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606037 MIM: 617015 | |||||||||||||||||||||||
Literature Links: |
CD96 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CD96 - CD96 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLCXD2 - phosphatidylinositol specific phospholipase C X domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185106.1 | 691 | Missense Mutation | CCT,GCT | P,A 41 | NP_001172035.1 | |
NM_153268.3 | 691 | Missense Mutation | CCT,GCT | P,A 41 | NP_695000.1 |
PLCXD2-AS1 - PLCXD2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |