Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGACCTTTTCATCAGATCCCC[A/G]CTCCAGCCCCTCGGAGAAGGAAACA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 147561 | |||||||||||||||||||||||
Literature Links: |
ITGB5 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ITGB5 - integrin subunit beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002213.4 | 2502 | UTR 3 | NP_002204.2 | |||
XM_005247436.2 | 2502 | UTR 3 | XP_005247493.1 | |||
XM_006713630.2 | 2502 | UTR 3 | XP_006713693.1 | |||
XM_017006351.1 | 2502 | UTR 3 | XP_016861840.1 | |||
XM_017006352.1 | 2502 | UTR 3 | XP_016861841.1 | |||
XM_017006353.1 | 2502 | UTR 3 | XP_016861842.1 | |||
XM_017006354.1 | 2502 | UTR 3 | XP_016861843.1 | |||
XM_017006355.1 | 2502 | UTR 3 | XP_016861844.1 |