Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAGTGTCTGAGGCTGAGCTGA[A/G]ACTGGAGGTGCTGAAGTGACTTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606173 MIM: 608790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC96 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC96 - coiled-coil domain containing 96 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRPEL1 - GrpE like 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100129931 - uncharacterized LOC100129931 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TADA2B - transcriptional adaptor 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152293.2 | Intron | NP_689506.2 | ||||
XM_011513595.1 | Intron | XP_011511897.1 |