Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGAAAGATATAAAGTGCTCAGC[G/T]TATTTTGCTGATTTTGAAAATCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602710 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APBB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSUN7 - NOP2/Sun RNA methyltransferase family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017008611.1 | 8219 | Intron | XP_016864100.1 | |||
XM_017008612.1 | 8219 | Intron | XP_016864101.1 | |||
XM_017008613.1 | 8219 | Intron | XP_016864102.1 | |||
XM_017008614.1 | 8219 | Intron | XP_016864103.1 | |||
XM_017008615.1 | 8219 | Intron | XP_016864104.1 |