Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGGGTTTCTACTCACTTGTTCA[C/T]ATAAACCAGACCCCATCATTCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF518B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF518B - zinc finger protein 518B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053042.2 | 8198 | UTR 3 | NP_444270.2 | |||
XM_005248193.2 | 8198 | UTR 3 | XP_005248250.1 | |||
XM_017008784.1 | 8198 | UTR 3 | XP_016864273.1 | |||
XM_017008785.1 | 8198 | UTR 3 | XP_016864274.1 | |||
XM_017008786.1 | 8198 | UTR 3 | XP_016864275.1 |