Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATGAGCTTGGCCACATCCACCA[C/T]CTGCCCAGGGGGAAGCAGAACCGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601481 | ||||||||||||||||||||
Literature Links: |
MYO10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO10 - myosin X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012334.2 | 5438 | Missense Mutation | ATG,GTG | M,V 2026 | NP_036466.2 | |
XM_005248306.4 | 5438 | Missense Mutation | ATG,GTG | M,V 1384 | XP_005248363.1 | |
XM_005248307.2 | 5438 | Missense Mutation | ATG,GTG | M,V 1383 | XP_005248364.1 | |
XM_006714475.2 | 5438 | Missense Mutation | ATG,GTG | M,V 2003 | XP_006714538.1 | |
XM_011514046.2 | 5438 | Missense Mutation | ATG,GTG | M,V 1383 | XP_011512348.1 |