Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTATTCTCTGAGCTAGGGAGAT[A/C]CTTTCCCTGAAGAATGGAGTTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612471 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGXT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGXT2 - alanine--glyoxylate aminotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306173.1 | 2043 | Intron | NP_001293102.1 | |||
NM_031900.3 | 2043 | UTR 3 | NP_114106.1 | |||
XM_005248337.2 | 2043 | UTR 3 | XP_005248394.1 | |||
XM_005248338.2 | 2043 | UTR 3 | XP_005248395.1 | |||
XM_017009748.1 | 2043 | UTR 3 | XP_016865237.1 |