Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGCCACTCCAGTTTATTGAAA[A/T]GAGTAAATTTATAGCTTTATTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120940 MIM: 601236 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9 - complement component 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DAB2 - DAB2, clathrin adaptor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244871.1 | 4509 | UTR 3 | NP_001231800.1 | |||
NM_001343.3 | 4509 | UTR 3 | NP_001334.2 |