Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAAGACAGCTGAGAACTTCCTC[C/T]TCTGAGCACTCAAACTTTTCACATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602739 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKAA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRKAA1 - protein kinase AMP-activated catalytic subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006251.5 | 1907 | Silent Mutation | GAA,GAG | E,E 315 | NP_006242.5 | |
NM_206907.3 | 1907 | Silent Mutation | GAA,GAG | E,E 330 | NP_996790.3 | |
XM_006714482.1 | 1907 | Silent Mutation | GAA,GAG | E,E 57 | XP_006714545.1 | |
XM_017009624.1 | 1907 | Silent Mutation | GAA,GAG | E,E 304 | XP_016865113.1 | |
XM_017009625.1 | 1907 | Silent Mutation | GAA,GAG | E,E 289 | XP_016865114.1 |
TTC33 - tetratricopeptide repeat domain 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |