Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGCCTAACTGTACAATGTTTA[C/T]GAAGGCTATAGCAGTGTATAGTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610103 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100Z PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
S100Z - S100 calcium binding protein Z | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130772.3 | 86 | Intron | NP_570128.2 | |||
XM_011543239.2 | 86 | Intron | XP_011541541.1 | |||
XM_011543240.2 | 86 | Intron | XP_011541542.1 | |||
XM_011543241.2 | 86 | Intron | XP_011541543.1 | |||
XM_011543244.2 | 86 | Intron | XP_011541546.1 | |||
XM_011543245.2 | 86 | Intron | XP_011541547.1 | |||
XM_017009171.1 | 86 | Intron | XP_016864660.1 | |||
XM_017009172.1 | 86 | UTR 5 | XP_016864661.1 | |||
XM_017009173.1 | 86 | Intron | XP_016864662.1 |