Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAGATATGAAGGAACTTGAAAGC[C/T]GTAAGTATATGAAGAAGGCAATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERINC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERINC5 - serine incorporator 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174071.2 | Intron | NP_001167542.1 | ||||
NM_001174072.2 | Intron | NP_001167543.1 | ||||
NM_178276.6 | Intron | NP_840060.1 | ||||
XM_011543304.1 | Intron | XP_011541606.1 | ||||
XM_017009323.1 | Intron | XP_016864812.1 | ||||
XM_017009324.1 | Intron | XP_016864813.1 | ||||
XM_017009325.1 | Intron | XP_016864814.1 | ||||
XM_017009326.1 | Intron | XP_016864815.1 |