Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGGCTGCAGCGGCCGCAGCGGC[C/T]GCAGCGCCCCCAGCGCCCCCAGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 126060 MIM: 600887 | ||||||||||||||||||||
Literature Links: |
DHFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHFR - dihydrofolate reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000791.3 | 260 | UTR 5 | NP_000782.1 | |||
NM_001290354.1 | 260 | UTR 5 | NP_001277283.1 | |||
NM_001290357.1 | 260 | UTR 5 | NP_001277286.1 |
MSH3 - mutS homolog 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002439.4 | 260 | Silent Mutation | GCC,GCT | A,A 60 | NP_002430.3 |
MTRNR2L2 - MT-RNR2-like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |