Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCACCTAGCTGGGACCTGTGGG[A/G]ACAGGAAGGAAATGTGGCAAGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL19A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL19A1 - collagen type XIX alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001858.5 | Intron | NP_001849.2 | ||||
XM_011535437.2 | Intron | XP_011533739.1 | ||||
XM_011535438.2 | Intron | XP_011533740.1 | ||||
XM_017010253.1 | Intron | XP_016865742.1 | ||||
XM_017010254.1 | Intron | XP_016865743.1 | ||||
XM_017010255.1 | Intron | XP_016865744.1 | ||||
XM_017010256.1 | Intron | XP_016865745.1 | ||||
XM_017010257.1 | Intron | XP_016865746.1 | ||||
XM_017010258.1 | Intron | XP_016865747.1 | ||||
XM_017010259.1 | Intron | XP_016865748.1 |