Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGCTGATCACCTAATGACTAAC[A/G]GTCATTTTCATCCTGGAAAATGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147460 MIM: 605442 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SOD2 - superoxide dismutase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000636.3 | Intron | NP_000627.2 | ||||
NM_001024465.2 | Intron | NP_001019636.1 | ||||
NM_001024466.2 | Intron | NP_001019637.1 | ||||
NM_001322814.1 | Intron | NP_001309743.1 | ||||
NM_001322815.1 | Intron | NP_001309744.1 | ||||
NM_001322816.1 | Intron | NP_001309745.1 | ||||
NM_001322817.1 | Intron | NP_001309746.1 | ||||
NM_001322819.1 | Intron | NP_001309748.1 | ||||
NM_001322820.1 | Intron | NP_001309749.1 |
WTAP - Wilms tumor 1 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270531.1 | Intron | NP_001257460.1 | ||||
NM_001270532.1 | Intron | NP_001257461.1 | ||||
NM_001270533.1 | Intron | NP_001257462.1 | ||||
NM_004906.4 | Intron | NP_004897.2 | ||||
NM_152857.2 | Intron | NP_690596.1 | ||||
NM_152858.2 | Intron | NP_690597.1 | ||||
XM_017011514.1 | Intron | XP_016867003.1 | ||||
XM_017011515.1 | Intron | XP_016867004.1 | ||||
XM_017011516.1 | Intron | XP_016867005.1 |