Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGCCCCGAACTCCATGGGTTTC[C/T]AGTAGAATTTCAAGCCAGATGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HFE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HFE - hemochromatosis | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000410.3 | 388 | Silent Mutation | TCC,TCT | S,S 76 | NP_000401.1 | |
NM_001300749.1 | 388 | Silent Mutation | TCC,TCT | S,S 76 | NP_001287678.1 | |
NM_139003.2 | 388 | Silent Mutation | TCC,TCT | S,S 76 | NP_620572.1 | |
NM_139004.2 | 388 | Silent Mutation | TCC,TCT | S,S 76 | NP_620573.1 | |
NM_139006.2 | 388 | Silent Mutation | TCC,TCT | S,S 76 | NP_620575.1 | |
NM_139007.2 | 388 | Intron | NP_620576.1 | |||
NM_139008.2 | 388 | Intron | NP_620577.1 | |||
NM_139009.2 | 388 | Silent Mutation | TCC,TCT | S,S 53 | NP_620578.1 | |
NM_139010.2 | 388 | Intron | NP_620579.1 | |||
NM_139011.2 | 388 | Intron | NP_620580.1 | |||
XM_011514543.2 | 388 | Silent Mutation | TCC,TCT | S,S 76 | XP_011512845.1 |