Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTAACTTGCCAAGTGTGGTTGC[C/T]TAGAAGCAAGAGGAAGAGCAAGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612217 | ||||||||||||||||||||
Literature Links: |
C6orf106 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf106 - chromosome 6 open reading frame 106 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022758.5 | 3234 | UTR 3 | NP_073595.2 | |||
NM_024294.3 | 3234 | UTR 3 | NP_077270.1 | |||
XM_005249298.2 | 3234 | Intron | XP_005249355.1 |