Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCATTCAGTGATTTTTGCAGAGT[A/C]TGGGTCTGGATTTTCTTTCCTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPARD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPARD - peroxisome proliferator activated receptor delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171818.1 | Intron | NP_001165289.1 | ||||
NM_001171819.1 | Intron | NP_001165290.1 | ||||
NM_001171820.1 | Intron | NP_001165291.1 | ||||
NM_006238.4 | Intron | NP_006229.1 | ||||
NM_177435.2 | Intron | NP_803184.1 | ||||
XM_005249193.1 | Intron | XP_005249250.1 | ||||
XM_006715120.1 | Intron | XP_006715183.1 | ||||
XM_006715123.1 | Intron | XP_006715186.1 | ||||
XM_011514707.1 | Intron | XP_011513009.1 | ||||
XM_011514709.1 | Intron | XP_011513011.1 | ||||
XM_011514710.1 | Intron | XP_011513012.1 | ||||
XM_017010971.1 | Intron | XP_016866460.1 | ||||
XM_017010972.1 | Intron | XP_016866461.1 | ||||
XM_017010973.1 | Intron | XP_016866462.1 | ||||
XM_017010974.1 | Intron | XP_016866463.1 | ||||
XM_017010975.1 | Intron | XP_016866464.1 |