Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGTCCCACTGCATTTCCAATTA[C/T]TAACAATTATTTTCACTTACAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143170 MIM: 601498 MIM: 601646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEA1 - male-enhanced antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318942.1 | 550 | Intron | NP_001305871.1 | |||
NM_001318943.1 | 550 | Intron | NP_001305872.1 | |||
NM_014623.3 | 550 | Intron | NP_055438.1 | |||
XM_005249121.3 | 550 | Intron | XP_005249178.1 | |||
XM_017010868.1 | 550 | UTR 3 | XP_016866357.1 |
PEX6 - peroxisomal biogenesis factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000287.3 | 550 | Intron | NP_000278.3 | |||
NM_001316313.1 | 550 | Intron | NP_001303242.1 |
PPP2R5D - protein phosphatase 2 regulatory subunit B'delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |