Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTACAGGGATATCAGCTTTGGGCA[A/G]AAGCAGAAGGCAGCTTCACGTAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609577 MIM: 611822 | ||||||||||||||||||||
Literature Links: |
CUL7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL7 - cullin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168370.1 | 890 | Intron | NP_001161842.1 | |||
NM_014780.4 | 890 | Intron | NP_055595.2 | |||
XM_005249503.2 | 890 | Intron | XP_005249560.1 | |||
XM_006715285.1 | 890 | Intron | XP_006715348.1 | |||
XM_011515019.2 | 890 | Intron | XP_011513321.1 | |||
XM_011515020.2 | 890 | Intron | XP_011513322.1 | |||
XM_011515021.1 | 890 | Intron | XP_011513323.1 | |||
XM_017011533.1 | 890 | Intron | XP_016867022.1 | |||
XM_017011534.1 | 890 | Intron | XP_016867023.1 | |||
XM_017011535.1 | 890 | Intron | XP_016867024.1 | |||
XM_017011536.1 | 890 | Intron | XP_016867025.1 | |||
XM_017011537.1 | 890 | Intron | XP_016867026.1 | |||
XM_017011538.1 | 890 | Intron | XP_016867027.1 | |||
XM_017011539.1 | 890 | Intron | XP_016867028.1 | |||
XM_017011540.1 | 890 | Intron | XP_016867029.1 |
KLC4 - kinesin light chain 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL2 - mitochondrial ribosomal protein L2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300848.1 | 890 | UTR 3 | NP_001287777.1 | |||
NM_015950.4 | 890 | Missense Mutation | TCT,TTT | S,F 302 | NP_057034.2 | |
XM_005249161.4 | 890 | Missense Mutation | TCT,TTT | S,F 270 | XP_005249218.1 |