Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAATAGCTGAGGTCTGTTGCCCA[A/G]AGTTATTAATTCCATTCATTTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRF5 - adhesion G protein-coupled receptor F5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098518.1 | Intron | NP_001091988.1 | ||||
NM_015234.4 | Intron | NP_056049.4 | ||||
XM_005248892.2 | Intron | XP_005248949.1 | ||||
XM_005248893.3 | Intron | XP_005248950.1 | ||||
XM_005248894.3 | Intron | XP_005248951.1 | ||||
XM_005248895.3 | Intron | XP_005248952.1 | ||||
XM_011514353.2 | Intron | XP_011512655.1 | ||||
XM_011514354.2 | Intron | XP_011512656.1 | ||||
XM_017010418.1 | Intron | XP_016865907.1 | ||||
XM_017010419.1 | Intron | XP_016865908.1 | ||||
XM_017010420.1 | Intron | XP_016865909.1 | ||||
XM_017010421.1 | Intron | XP_016865910.1 | ||||
XM_017010422.1 | Intron | XP_016865911.1 | ||||
XM_017010423.1 | Intron | XP_016865912.1 | ||||
XM_017010424.1 | Intron | XP_016865913.1 |
MEP1A - meprin A subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |