Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAATTTCTCCTAACACTAAGTTA[A/G]AAAATCATTTGCATCATGCTGTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611533 MIM: 603854 MIM: 604483 | ||||||||||||||||||||
Literature Links: |
NOL7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOL7 - nucleolar protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317724.1 | 1892 | Intron | NP_001304653.1 | |||
NM_016167.4 | 1892 | Intron | NP_057251.2 |
RANBP9 - RAN binding protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005493.2 | 1892 | UTR 3 | NP_005484.2 | |||
XM_006714945.2 | 1892 | UTR 3 | XP_006715008.1 | |||
XM_011514205.2 | 1892 | UTR 3 | XP_011512507.1 | |||
XM_017010149.1 | 1892 | UTR 3 | XP_016865638.1 |
SIRT5 - sirtuin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |