Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTCAGTTGATACTAACTAAAAC[C/T]AAAAAAGGCACCAGGTTTTCATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611533 MIM: 603854 MIM: 604483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOL7 - nucleolar protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317724.1 | Intron | NP_001304653.1 | ||||
NM_016167.4 | Intron | NP_057251.2 |
RANBP9 - RAN binding protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005493.2 | Intron | NP_005484.2 | ||||
XM_006714945.2 | Intron | XP_006715008.1 | ||||
XM_011514205.2 | Intron | XP_011512507.1 | ||||
XM_017010149.1 | Intron | XP_016865638.1 |
SIRT5 - sirtuin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |