Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGATGAAACATGAAGACTTTA[C/T]GCTAAGTGAATAATTCAAGGATGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCB5 - ATP binding cassette subfamily B member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163941.1 | Intron | NP_001157413.1 | ||||
NM_001163942.1 | Intron | NP_001157414.1 | ||||
NM_001163993.2 | Intron | NP_001157465.1 | ||||
NM_178559.5 | Intron | NP_848654.3 | ||||
XM_011515367.2 | Intron | XP_011513669.1 |