Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGAGAACATCCCAGGAAAACCTA[C/T]AGTCCACTACTGGACATGCCTGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREB5 - cAMP responsive element binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011666.2 | Intron | NP_001011666.1 | ||||
NM_004904.3 | Intron | NP_004895.2 | ||||
NM_182898.3 | Intron | NP_878901.2 | ||||
NM_182899.4 | Intron | NP_878902.2 | ||||
XM_005249906.1 | Intron | XP_005249963.1 | ||||
XM_011515618.2 | Intron | XP_011513920.1 | ||||
XM_017012806.1 | Intron | XP_016868295.1 | ||||
XM_017012807.1 | Intron | XP_016868296.1 | ||||
XM_017012808.1 | Intron | XP_016868297.1 | ||||
XM_017012809.1 | Intron | XP_016868298.1 | ||||
XM_017012810.1 | Intron | XP_016868299.1 |