Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTATATGTTGACAGGGTGGTGA[A/C]TTCACACGCCATAATGGCACTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123840 | ||||||||||||||||||||
Literature Links: |
PPIA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPIA - peptidylprolyl isomerase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300981.1 | 287 | Intron | NP_001287910.1 | |||
NM_021130.4 | 287 | Missense Mutation | GAA,GAC | E,D 66 | NP_066953.1 |