Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTCCTGGGGTGGGGGTCGGGGG[C/T]ACTTTGGTTCCCAGGGTTTGGAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUB1 - negative regulator of ubiquitin like proteins 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR86 - WD repeat domain 86 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284260.1 | 1810 | Intron | NP_001271189.1 | |||
NM_001284261.1 | 1810 | Intron | NP_001271190.1 | |||
NM_001284262.1 | 1810 | Intron | NP_001271191.1 | |||
NM_198285.2 | 1810 | Intron | NP_938026.2 | |||
XM_005249989.3 | 1810 | Intron | XP_005250046.1 | |||
XM_005249990.4 | 1810 | Intron | XP_005250047.1 | |||
XM_006715966.2 | 1810 | Intron | XP_006716029.1 | |||
XM_011516144.2 | 1810 | UTR 3 | XP_011514446.1 | |||
XM_011516145.2 | 1810 | UTR 3 | XP_011514447.1 | |||
XM_011516146.2 | 1810 | Intron | XP_011514448.1 | |||
XM_011516147.2 | 1810 | UTR 3 | XP_011514449.1 | |||
XM_011516148.1 | 1810 | Intron | XP_011514450.1 | |||
XM_011516150.1 | 1810 | Intron | XP_011514452.1 | |||
XM_011516151.2 | 1810 | UTR 3 | XP_011514453.1 | |||
XM_011516152.1 | 1810 | Intron | XP_011514454.1 |