Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCAGATTGGACATTTGAGTGCA[C/T]TTTACCTTGTGTGATGAGACAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB19 - RAB19, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008749.2 | Intron | NP_001008749.2 | ||||
XM_005249996.3 | Intron | XP_005250053.1 | ||||
XM_005249997.3 | Intron | XP_005250054.1 | ||||
XM_005249998.3 | Intron | XP_005250055.1 | ||||
XM_017012207.1 | Intron | XP_016867696.1 |
SLC37A3 - solute carrier family 37 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287498.1 | Intron | NP_001274427.1 | ||||
NM_032295.3 | Intron | NP_115671.1 | ||||
NM_207113.2 | Intron | NP_996996.1 | ||||
XM_011516626.2 | Intron | XP_011514928.1 | ||||
XM_011516627.2 | Intron | XP_011514929.1 | ||||
XM_017012712.1 | Intron | XP_016868201.1 | ||||
XM_017012713.1 | Intron | XP_016868202.1 | ||||
XM_017012714.1 | Intron | XP_016868203.1 | ||||
XM_017012715.1 | Intron | XP_016868204.1 | ||||
XM_017012716.1 | Intron | XP_016868205.1 | ||||
XM_017012717.1 | Intron | XP_016868206.1 | ||||
XM_017012718.1 | Intron | XP_016868207.1 |