Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTACCCTGTACTTGCCCCCACCC[A/C]AAACAAAAACAACAACAAAGAAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605568 | ||||||||||||||||||||
Literature Links: |
LOC101927550 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927550 - uncharacterized LOC101927550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMURF1 - SMAD specific E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199847.1 | 3875 | UTR 3 | NP_001186776.1 | |||
NM_020429.2 | 3875 | UTR 3 | NP_065162.1 | |||
NM_181349.2 | 3875 | UTR 3 | NP_851994.1 | |||
XM_017012457.1 | 3875 | UTR 3 | XP_016867946.1 |