Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCGAGGTACGCGGTGCGGTCTC[A/C]CGGTACCCGGAGCGGCTCTGATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601260 MIM: 601261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZKSCAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZKSCAN1 - zinc finger with KRAB and SCAN domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN21 - zinc finger and SCAN domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145914.2 | 21 | UTR 5 | NP_666019.1 | |||
XM_005250568.4 | 21 | Intron | XP_005250625.1 | |||
XM_006716113.3 | 21 | UTR 5 | XP_006716176.1 | |||
XM_017012584.1 | 21 | Intron | XP_016868073.1 | |||
XM_017012585.1 | 21 | UTR 5 | XP_016868074.1 |