Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAATCGAATATCTCATGGCCTAC[A/C]GGGACTTTCTGCAGTGCCTCTGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 238331 | ||||||||||||||||||||
Literature Links: |
DLD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLD - dihydrolipoamide dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000108.4 | 218 | Missense Mutation | CAG,CCG | Q,P 25 | NP_000099.2 | |
NM_001289750.1 | 218 | UTR 5 | NP_001276679.1 | |||
NM_001289751.1 | 218 | Missense Mutation | CAG,CCG | Q,P 25 | NP_001276680.1 | |
NM_001289752.1 | 218 | Missense Mutation | CAG,CCG | Q,P 25 | NP_001276681.1 |