Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAGATGGGCTCGGTTTTAACTA[C/T]GTCCGAGGAAGCCTGGAAAAAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTTNBP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTTNBP2 - cortactin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033427.2 | Intron | NP_219499.1 | ||||
XM_005250634.4 | Intron | XP_005250691.1 | ||||
XM_005250635.3 | Intron | XP_005250692.1 | ||||
XM_011516615.2 | Intron | XP_011514917.1 | ||||
XM_011516616.2 | Intron | XP_011514918.1 | ||||
XM_011516618.2 | Intron | XP_011514920.1 | ||||
XM_017012706.1 | Intron | XP_016868195.1 | ||||
XM_017012707.1 | Intron | XP_016868196.1 | ||||
XM_017012708.1 | Intron | XP_016868197.1 |