Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCCACATCACTATTTTTGCAA[C/T]CAAGAGAGACCACAATTGGGAAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189964 MIM: 601558 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2E2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2E2 - general transcription factor IIE subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002095.4 | Intron | NP_002086.1 | ||||
XM_011544509.2 | Intron | XP_011542811.1 | ||||
XM_011544510.1 | Intron | XP_011542812.1 | ||||
XM_017013363.1 | Intron | XP_016868852.1 | ||||
XM_017013364.1 | Intron | XP_016868853.1 |
RBPMS - RNA binding protein with multiple splicing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |