Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGCCTCTGGTTGCTGCTGGCCGG[A/G]CTCTGCGGCCTCCTGGCGTCAAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM32 - ADAM metallopeptidase domain 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001313994.1 | 71 | Missense Mutation | GAC,GGC | D,G 44 | NP_001300923.1 | |
NM_145004.6 | 71 | Silent Mutation | GGA,GGG | G,G 10 | NP_659441.3 | |
XM_006716298.2 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_006716361.1 | |
XM_006716299.1 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_006716362.1 | |
XM_011544433.1 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_011542735.1 | |
XM_011544434.1 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_011542736.1 | |
XM_017013183.1 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_016868672.1 | |
XM_017013184.1 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_016868673.1 | |
XM_017013185.1 | 71 | Intron | XP_016868674.1 | |||
XM_017013186.1 | 71 | Silent Mutation | GGA,GGG | G,G 10 | XP_016868675.1 | |
XM_017013187.1 | 71 | Intron | XP_016868676.1 | |||
XM_017013188.1 | 71 | Intron | XP_016868677.1 |
ADAM9 - ADAM metallopeptidase domain 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |