Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTTATGTTTATGCTTCTTCTT[C/T]TTTTTCTTGTGCTCATGGTGATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604912 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF2 - TATA-box binding protein associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003184.3 | 3607 | Silent Mutation | AAA,AAG | K,K 1157 | NP_003175.1 | |
XM_006716621.3 | 3607 | Silent Mutation | AAA,AAG | K,K 1209 | XP_006716684.1 | |
XM_011517259.2 | 3607 | Silent Mutation | AAA,AAG | K,K 1172 | XP_011515561.1 | |
XM_017013779.1 | 3607 | Silent Mutation | AAA,AAG | K,K 1120 | XP_016869268.1 | |
XM_017013780.1 | 3607 | Silent Mutation | AAA,AAG | K,K 1040 | XP_016869269.1 | |
XM_017013781.1 | 3607 | Silent Mutation | AAA,AAG | K,K 1040 | XP_016869270.1 | |
XM_017013782.1 | 3607 | Silent Mutation | AAA,AAG | K,K 1040 | XP_016869271.1 | |
XM_017013783.1 | 3607 | Silent Mutation | AAA,AAG | K,K 998 | XP_016869272.1 | |
XM_017013784.1 | 3607 | Silent Mutation | AAA,AAG | K,K 988 | XP_016869273.1 | |
XM_017013785.1 | 3607 | Silent Mutation | AAA,AAG | K,K 988 | XP_016869274.1 | |
XM_017013786.1 | 3607 | Silent Mutation | AAA,AAG | K,K 988 | XP_016869275.1 | |
XM_017013787.1 | 3607 | Silent Mutation | AAA,AAG | K,K 951 | XP_016869276.1 | |
XM_017013788.1 | 3607 | Silent Mutation | AAA,AAG | K,K 792 | XP_016869277.1 | |
XM_017013789.1 | 3607 | Silent Mutation | AAA,AAG | K,K 740 | XP_016869278.1 | |
XM_017013790.1 | 3607 | Silent Mutation | AAA,AAG | K,K 740 | XP_016869279.1 | |
XM_017013791.1 | 3607 | Silent Mutation | AAA,AAG | K,K 710 | XP_016869280.1 |