Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGAGTTGAGACCTTAGAGCTG[C/T]AAAAATGAGTGAAGGATTAGAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610931 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFAT - zinc finger and AT-hook domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029939.3 | Intron | NP_001025110.2 | ||||
NM_001167583.2 | Intron | NP_001161055.1 | ||||
NM_001174157.1 | Intron | NP_001167628.1 | ||||
NM_001174158.1 | Intron | NP_001167629.1 | ||||
NM_001289394.1 | Intron | NP_001276323.1 | ||||
NM_020863.3 | Intron | NP_065914.2 | ||||
XM_011517203.1 | Intron | XP_011515505.1 | ||||
XM_011517204.2 | Intron | XP_011515506.1 | ||||
XM_011517206.1 | Intron | XP_011515508.1 | ||||
XM_017013716.1 | Intron | XP_016869205.1 |